missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine pHGF Protein

A cDNA sequence encoding the pHGF was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13039-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15972319

  • 131.00 CHF / 2 Mikrogramm
Verfügbar ab: 19-09-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

pHGF Protein
Research Use Only
Recombinant Protein
Porcine
The sterile protein powder is lyophilized with no additives.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Porcine
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote