missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine pHGF Protein
A cDNA sequence encoding the pHGF was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 4340.00 CHF
Spezifikation
Name | pHGF Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Porcine |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15972319
|
enQuireBio™
QP13039-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15952319
|
enQuireBio™
QP13039-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15962319
|
enQuireBio™
QP13039-1MG |
1 mg |
4340.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
pHGF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Porcine | |
The sterile protein powder is lyophilized with no additives. |