missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine pHGF Protein
A cDNA sequence encoding the pHGF was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13039-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
pHGF Protein | |
Research Use Only | |
Recombinant Protein | |
Porcine | |
The sterile protein powder is lyophilized with no additives. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |