missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine IL 2 Protein
A cDNA sequence encoding the IL 2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10699-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
396868 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.5ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Porcine | |
Untagged | |
The protein (1 mg/ml) was lyophilized was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |
IL 2 Protein | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKNT KKQLEPLLLD LQLLLKEVKN YENADLSRML TFKFYMPKQA TELKHLQCLV EELKALEGVL NLGQSKNSDS ANIKESMNNI NVTVLELKGS ETSFKCEYDD ETVTAVEFLN KWITFCQSIY STLT | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |