missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine IL 2 Protein
A cDNA sequence encoding the IL 2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 6775.00 CHF
Spezifikation
Gen-ID (Entrez) | 396868 |
---|---|
Name | IL 2 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gensymbol | IL2 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15984688
|
enQuireBio™
QP10699-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964688
|
enQuireBio™
QP10699-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974688
|
enQuireBio™
QP10699-1MG |
1 mg |
6775.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
396868 | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKNT KKQLEPLLLD LQLLLKEVKN YENADLSRML TFKFYMPKQA TELKHLQCLV EELKALEGVL NLGQSKNSDS ANIKESMNNI NVTVLELKGS ETSFKCEYDD ETVTAVEFLN KWITFCQSIY STLT | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.5ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Porcine | |
Untagged | |
The protein (1 mg/ml) was lyophilized was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |