missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Y. Enterocolitica (O:9) LcrV Protein
A cDNA sequence encoding the Y. Enterocolitica (O:9) LcrV was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13975-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Y. Enterocolitica (O:9) LcrV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Y. Enterocolitica (O:9) LcrV is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |