missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Luciferase Firefly Protein
A cDNA sequence encoding the Luciferase Firefly was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10776-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Luciferase Firefly Protein | |
Research Use Only | |
Specific activity is >1x109 light units/mg. One luciferase enzyme units will produce one Relative Light Unit (RLU) at pH7.5 at 25C. | |
Other | |
His | |
The Luciferase solution (0.5 mg/ml) contains 20mM Tris-HCl (pH 8.0), 1mM DTT and 10% glycerol. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTSKVY DPEQRKRMIT GPQWWARCKQ MNVLDSFINY YDSEKHAENA VIFLHGNAAS SYLWRHVVPH IEPVARCIIP DLIGMGKSGK SGNGSYRLLD HYKYLTAWFE LLNLPKKIIF VGHDWGACLA FHYSYEHQDK IKAIVHAESV VDVIESWDEW PDIEEDIALI KSEEGEKMVL ENNFFVETML PSKIMRKLEP EEFAAYLEPF KEKGEVRRPT LSWPREIPLV KGGKPDVVQI VRNYNAYLRA SDDLPKMFIE SDPGFFSNAI VEGAKKFPNT EFVKVKGLHF SQEDAPDEMG KYIKSFVERV LKNEQ | |
Greater than 95.0% as determined by SDS-PAGE. |