Learn More
enQuireBio™ Recombinant other Alarelin Protein
Recombinant Protein
Marke: enQuireBio™ QP10997-10mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Alarelin Protein | |
Research Use Only | |
Recombinant Protein | |
Untagged | |
The Alarelin was lyophilized with no additives. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
5-oxo-pro-His-Trp-Ser-Tyr-D-Ala-Leu-Arg-Pro-Nhet x CH3COOH | |
Greater than 98.0% as determined by Analysis by RP-HPLC. |