missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SF20 Protein
A cDNA sequence encoding the SF20 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13475-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SF20 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
Mouse SF20 was lyophilized from a 0.2microm filtered concentrated (1.0 mg/ml) solution in PBS, pH 7.4. |