missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse REN Protein
A cDNA sequence encoding the REN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13297-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
REN Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LPTRTATFER IPLKKMPSVR EILEERGVDM TRLSAEWGVF TKRPSLTNLT SPVVLTNYLN TQYYGEIGIG TPPQTFKVIF DTGSANLWVP STKCSRLYLA CGIHSLYESS DSSSYMENGS DFTIHYGSGR VKGFLSQDSV TVGGITVTQT FGEVTELPLI PFMLAKFDGV LGMGFPAQAV GGVTPVFDHI LSQGVLKEEV FSVYYNRGSH LLGGEVVLGG SDPQHYQGNF HYVSISKTDS WQITMKGVSV GSSTLLCEEG CAVVVDTGSS FISAPTSSLK LIMQALGAKE KRIEEYVVNC SQVPTLPDIS FDLGGRAYTL SSTDYVLQYP NRRDKLCTLA LHAMDIPPPT GPVWVLGATF IRKFYTEFDR HNNRIGFALA RHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
REN protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |