missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Procalcitonin Protein
A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13152-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Procalcitonin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVPLRSILESS PGMATLSEEE VRLLAALVQD YMQMKARELE QEEEQEAEGS SLDSPRSKRC GNLSTCMLGT YTQDLNKFHT FPQTSIGVEA PGKKRDVAKD LETNHQSHFG N | |
Greater than 94.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
Procalcitonin was filtered (0.4 microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5. |