missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRDX2 Protein
A cDNA sequence encoding the PRDX2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10500-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PRDX2 Protein | |
Research Use Only | |
Specific activity is > 700 pmol/min/ug, Activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for minute. | |
Mouse | |
His | |
PRDX2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol 1mM DTT. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMASGNA QIGKSAPDFT ATAVVDGAFK EIKLSDYRGK YVVLFFYPLD FTFVCPTEII AFSDHAEDFR KLGCEVLGVS VDSQFTHLAW INTPRKEGGL GPLNIPLLAD VTKSLSQNYG VLKNDEGIAY RGLFIIDAKG VLRQITVNDL PVGRSVDEAL RLVQAFQYTD EHGEVCPAGW KPGSDTIKPN VDDSKEYFSK HN | |
Greater than 90.0% as determined by SDS-PAGE. |