missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL5 Protein
A DNA sequence encoding the Mouse IL5, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10454-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_034688.1 | |
IL5 Protein | |
Research Use Only | |
IL5 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Mouse | |
Untagged | |
0.2 m filtered 20 mM Tris, pH 9.0, 150 mM NaCl, lyophilized |
16191 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MEIPMSTVVK ETLTQLSAHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEMLFQNL SLIKKYIDRQ KEKCGEERRR TRQFLDYLQE FLGVMSTEWA MEG | |
Greater than 98% as determined by SDS-PAGE and HPLC. |