missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL36B / IL1F8 Protein
A DNA sequence encoding the Mouse IL36B / IL1F8, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10436-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_081439.1 | |
IL36B / IL1F8 Protein | |
Research Use Only | |
Il36b | |
The specific activity determined by its ability in a functional ELISA. Immobilized rMuIL-36beta at 1 ug/mL can bind recombinant murine IL-1 Rrp2 with a range of 0.15-5 ug/mL. | |
Mouse | |
Untagged | |
0.2 um filtered 20 mM Tris, 300 mM NaCl, pH 8.0, 5 % trehalose, lyophilized |
69677 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MMAFPPQSCV HVLPPKSIQM WEPNHNTMHG SSQSPRNYRV HDSQQMVWVL TGNTLTAVPA SNNVKPVILS LIACRDTEFQ DVKKGNLVFL GIKNRNLCFC CVEMEGKPTL QLKEVDIMNL YKERKAQKAF LFYHGIEGST SVFQSVLYPG WFIATSSIER QTIILTHQRG KLVNTNFYIE SEK | |
Greater than 95% as determined by SDS-PAGE and HPLC. |