missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL17RB Protein
A cDNA sequence encoding the IL17RB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12410-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
192199 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPREPTIQC GSETGPSPEW MVQHTLTPGD LRDLQVELVK TSVAAEEFSI LMNISWILRA DASIRLLKAT KICVSGKNNM NSYSCVRCNY TEAFQSQTRP SGGKWTFSYV GFPVELSTLY LISAHNIPNA NMNEDSPSLS VNFTSPGCLN HVMKYKKQCT EAGSLWDPDI TACKKNEKMV EVNFTTNPLG NRYTILIQRD TTLGFSRVLE NKLMRTSVAI PVTEESEGAV VQLTPYLHTC GNDCIRREGT VVLCSETSAP IPPDDNRRML GGVEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
IL17RB Protein | |
Research Use Only | |
Rspo1 | |
Mouse | |
His | |
IL17RB protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |