missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL17RB Protein
A cDNA sequence encoding the IL17RB was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Gen-ID (Entrez) | 192199 |
---|---|
Name | IL17RB Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | Rspo1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15920339
|
enQuireBio™
QP12410-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15900339
|
enQuireBio™
QP12410-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15910339
|
enQuireBio™
QP12410-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 14-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
192199 | |
Research Use Only | |
Rspo1 | |
Mouse | |
His | |
IL17RB protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
IL17RB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPREPTIQC GSETGPSPEW MVQHTLTPGD LRDLQVELVK TSVAAEEFSI LMNISWILRA DASIRLLKAT KICVSGKNNM NSYSCVRCNY TEAFQSQTRP SGGKWTFSYV GFPVELSTLY LISAHNIPNA NMNEDSPSLS VNFTSPGCLN HVMKYKKQCT EAGSLWDPDI TACKKNEKMV EVNFTTNPLG NRYTILIQRD TTLGFSRVLE NKLMRTSVAI PVTEESEGAV VQLTPYLHTC GNDCIRREGT VVLCSETSAP IPPDDNRRML GGVEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |