Learn More
enQuireBio™ Recombinant Mouse IL-31 / IL31 Protein
Recombinant Protein
Marke: enQuireBio™ QP5477-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_083870.1 | |
IL-31 / IL31 Protein | |
Research Use Only | |
IL31 | |
Recombinant Protein | |
E. coli | |
MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C | |
Greater than 95% as determined by SDS-PAGE. |
76399 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL-31 / IL31 induces IL-8 production in human PBMCs which was detected via ELISA (typical ED50 is ? 25 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |