missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 12 Protein
A cDNA sequence encoding the IL 12 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10689-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
IL 12 Protein | |
Research Use Only | |
IL-12 Mouse has full biological activity when compared to standards. The ED50 is determined by the dose-dependent induced expression of CD25 on CD8+ mouse T cells is 10-20 ng/ml corresponding to a specific activity of 50,000-100,000units/mg. | |
Mouse | |
Untagged | |
The protein was lyophilized from 0.5xPBS, pH 7.5. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
p35 Subunit: RVIPVSGPAR CLSQSRNLLK TTDDMVKTAR EKLKHYSCTA EDIDHEDITR DQTSTLKTCL PLELHKNESC LATRETSSTT RGSCLPPQKT SLMMTLCLGS IYEDLKMYQT EFQAINAALQ NHNHQQIILD KGMLVAIDEL MQSLNHNGET LRQKPPVGEA DPYRVKMKLC ILLHAFSTRV VTINRVMGYL SSA p40 Subunit: MWELEKDVYV VEVDWTPDAP GETVNLTCDT PEEDDITWTS DQRHGVIGSG KTLTITVKEF LDAGQYTCHK GGETLSHSHL LLHKKENGIW STEILKNFKN KTFLKCEAPN YSGRFTCSWL VQRNMDLKFN IKSSSSSPDS RAVTCGMASL SAEKVTLDQR DYEKYSVSCQ EDVTCPTAEE TLPIELALEA RQQNKYENYS TSFFIRDIIK PDPPKNLQMK PLKNSQVEVS WEYPDSWSTP HSYFSLKFFV RIQRKKEKMK ETEEGCNQKG AFLVEKTSTE VQCKGGNVCV QAQDRYYNSS CSKWACVPCR VRS | |
Greater than 95% as determined by SDS-PAGE. |