missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-1 alpha / IL1A / IL1F1 Protein
A DNA sequence encoding the Mouse IL-1 alpha / IL1A / IL1F1 Protein, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10448-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_034684.2 | |
IL-1 alpha / IL1A / IL1F1 Protein | |
Research Use Only | |
IL1A | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 2 pg/ml, corresponding to a specific activity of > 5.0 M 108 IU/mg. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
16175 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SAPYTYQSDL RYKLMKLVRQ KFVMNDSLNQ TIYQDVDKHY LSTTWLNDLQ QEVKFDMYAY SSGGDDSKYP VTLKISDSQL FVSAQGEDQP VLLKELPETP KLITGSETDL IFFWKSINSK NYFTSAAYPE LFIATKEQSR VHLARGLPSM TDFQIS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |