missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL21a Protein
Recombinant Protein
Marke: enQuireBio™ QP5443-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001180596.1 | |
CCL21a Protein | |
Research Use Only | |
CCL21 | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
100504362 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SDGGGQDCCL KYSQKKIPYS IVRGYRKQEP SLGCPIPAIL FSPRKHSKPE LCANPEEGWV QNLMRRLDQP PAPGKQSPGC RKNRGTSKSG KKGKGSKGCK RTEQTQPSRG | |
Greater than 95% as determined by SDS-PAGE. |