missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL17 / TARC / SCYA17 Protein
A DNA sequence encoding the Mouse CCL17 / TARC / SCYA17, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10272-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_035462.2 | |
CCL17 / TARC / SCYA17 Protein | |
Research Use Only | |
CCL17 | |
The biologically active determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 1.0-10 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20295 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ARATNVGREC CLDYFKGAIP IRKLVSWYKT SVECSRDAIV FLTVQGKLIC ADPKDKHVKK AIRLVKNPRP | |
Greater than 97% as determined by SDS-PAGE and HPLC. |