missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL16 / Interleukin-16 Protein
A DNA sequence encoding the Rhesus Macaque IL16 / Interleukin-16, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10443-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001027980.1 | |
IL16 / Interleukin-16 Protein | |
Research Use Only | |
IL16 | |
The biological activity determined by a chemotaxis bioassay using human peripheral T lymphocytes is in a concentration range of 1.0-100 ng/ml. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
574100 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SAASASAASD VSVESSAEAT VYTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETIQPG DEILQLAGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QPKETTAAAD S | |
Greater than 98% as determined by SDS-PAGE and HPLC. |