missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Parainfluenza Type-3 Protein
A cDNA sequence encoding the Parainfluenza Type-3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12961-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Influenza Parainfluenza Type-3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Parainfluenza type-3 solution contains STE, 0.09% sodium azide (NaN3) and 0.005% thimerosal. Do not centrifuge! Mix thoroughly before use! |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |