missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YBX1 Protein
A cDNA sequence encoding the YBX1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13982-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
YBX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSEAET QQPPAAPPAA PALSAADTKP GTTGSGAGSG GPGGLTSAAP AGGDKKVIAT KVLGTVKWFN VRNGYGFINR NDTKEDVFVH QTAIKKNNPR KYLRSVGDGE TVEFDVVEGE KGAEAANVTG PGGVPVQGSK YAADRNHYRR YPRRRGPPRN YQQNYQNSES GEKNEGSESA PEGQAQQRRP YRRRRFPPYY MRRPYGRRPQ YSNPPVQGEV MEGADNQGAG EQGRPVRQNM YRGYRPRFRR GPPRQRQPRE DGNEEDKENQ GDETQGQQPP QRRYRRNFNY RRRRPENPKP QDGKETKAAD PPAENSSAPE AEQGGAE | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
YBX1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |