missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human YBX1 Protein

A cDNA sequence encoding the YBX1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13982-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965309

  • 3905.00 CHF / 1 Milligramm
Verfügbar ab: 27-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

YBX1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSEAET QQPPAAPPAA PALSAADTKP GTTGSGAGSG GPGGLTSAAP AGGDKKVIAT KVLGTVKWFN VRNGYGFINR NDTKEDVFVH QTAIKKNNPR KYLRSVGDGE TVEFDVVEGE KGAEAANVTG PGGVPVQGSK YAADRNHYRR YPRRRGPPRN YQQNYQNSES GEKNEGSESA PEGQAQQRRP YRRRRFPPYY MRRPYGRRPQ YSNPPVQGEV MEGADNQGAG EQGRPVRQNM YRGYRPRFRR GPPRQRQPRE DGNEEDKENQ GDETQGQQPP QRRYRRNFNY RRRRPENPKP QDGKETKAAD PPAENSSAPE AEQGGAE
Greater than 85.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
YBX1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 20% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote