missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human XPB GST (N-Term) Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
2 μg
Packungsgröße:
2 Mikrogramm
Beschreibung
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Zusammensetzung | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gen-ID (Entrez) | 2071 |
| Molekulargewicht | M.W. (Theoretical): 115.7 kDa |
| Menge | 2 μg |
| Lagerungsbedingungen | Store at -80°C. Avoid freeze-thaw cycles. |
| Gen-Alias | Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit |
| Gensymbol | ERCC3 |
| Forschungskategorie | Cancer, DNA Repair, Nucleotide Excision Repair |
| Reinheits- oder Qualitätsgrad | >80% by SDS-PAGE and Coomassie blue staining |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur