missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSR2 Protein
A cDNA sequence encoding the TSR2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13830-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
90121 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGAAED ARALFRAGVC AALEAWPALQ IAVENGFGGV HSQEKAKWLG GAVEDYFMRN ADLELDEVED FLGELLTNEF DTVVEDGSLP QVSQQLQTMF HHFQRGDGAA LREMASCITQ RKCKVTATAL KTARETDEDE DDVDSVEEME VTATNDGAAT DGVCPQPEPS DPDAQTIKEE DIVEDGWTIV RRK | |
Greater than 90% as determined by SDS-PAGE. |
TSR2 Protein | |
Research Use Only | |
TSR2 | |
Human | |
His | |
TSR2 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4) and 10% glycerol. |