missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRAIL R2 / CD262 / TNFRSF10B Protein
A DNA sequence encoding the Human TRAIL R2 / CD262 / TNFRSF10B, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10367-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_003833.4 | |
TRAIL R2 / CD262 / TNFRSF10B Protein | |
Research Use Only | |
TNFRSF10B | |
rHusTRAIL-R2 reduced the production of LPS- induced TNF by its ability to neutralize endogenous TRAIL in fresh human PBMC. In this assay, endogenous TRAIL is induced during a 24 hour exposure to LPS (10 ng/mL) but in the presence of rHusTRAIL-R2, TRAIL-induced TNF is suppressed. | |
Human | |
Untagged | |
0.2um filtered PBS, pH 7.4, lyophilized |
8795 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ESALITQQDL APQQRAAPQQ KRSSPSEGLC PPGHHISEDG RDCISCKYGQ DYSTHWNDLL FCLRCTRCDS GEVELSPCTT TRNTVCQCEE GTFREEDSPE MCRKCRTGCP RGMVKVGDCT PWSDIECVHK ES | |
Greater than 97% as determined by SDS-PAGE and HPLC. |