missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF18 Protein
A DNA sequence encoding the Human TNFSF18, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10373-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_005083.2 | |
TNFSF18 Protein | |
Research Use Only | |
TNFSF18 | |
The biological activity as determined by stimulation of IL-8 production using human PBMC is in a concentration range of 0.1-10 ng/ml. | |
Human | |
Untagged | |
0.2 m filtered PBS, pH 7.4, lyophilized |
8995 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ETAKEPCMAK FGPLPSKWQM ASSEPPCVNK VSDWKLEILQ NGLYLIYGQV APNANYNDVA PFEVRLYKNK DMIQTLTNKS KIQNVGGTYE LHVGDTIDLI FNSEHQVLKN NTYWGIILLA NPQFIS | |
Greater than 98% as determined by SDS-PAGE and HPLC. |