missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF17 / BCMA Protein
A DNA sequence encoding the Human TNFRSF17 / BCMA, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10365-ec-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001183.2 | |
TNFRSF17 / BCMA Protein | |
Research Use Only | |
TNFRSF17 | |
The ED50 as determined by its ability to inhibit APRIL-mediated proliferation of anti-IgM stimulated murine B cells is no less than 40 ng/ml, corresponding to a specific activity of > 2.5 M 104 IU/mg in the presence of 100 ng/ml human APRIL. | |
Human | |
Untagged | |
0.2 um filtered 30 % acetonitrile, 0.1 % TFA, lyophilized |
608 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AGQCSQNEYF DSLLHACIPC QLRCSSNTPP LTCQRYCNAS VTNSVKGTNA | |
Greater than 98% as determined by SDS-PAGE and HPLC. |