missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIR domain-containing adapter molecule 1 Protein
A DNA sequence encoding the Homo sapiens (Human) TIR domain-containing adapter molecule 1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10006-iv-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_891549.1 | |
TIR domain-containing adapter molecule 1 Protein | |
Research Use Only | |
TICAM1 | |
Recombinant Protein | |
E. coli | |
MACTGPSLPS AFDILGAAGQ DKLLYLKHKL KTPRPGCQGQ DLLHAMVLLK LGQETEARIS LEALKADAVA RLVARQWAGV DSTEDPEEPP DVSWAVARLY HLLAEEKLCP ASLRDVAYQE AVRTLSSRDD HRLGELQDEA RNRCGWDIAG DPGSIRTLQS NLGCLPPSSA LPSGTRSLPR PIDGVSDWSQ GCSLRSTGSP ASLASNLEIS QSPTMPFLSL HRSPHGPSKL CDDPQASLVP EPVPGGCQEP EEMSWPPSGE IASPPELPSS PPPGLPEVAP DATSTGLPDT PAAPETSTNY PVECTEGSAG PQSLPLPILE PVKNPCSVKD QTPLQLSVED TTSPNTKPCP PTPTTPETSP PPPPPPPSST PCSAHLTPSS LFPSSLESSS EQKFYNFVIL HARADEHIAL RVREKLEALG VPDGATFCED FQVPGRGELS CLQDAIDHSA FIILLLTSNF DCRLSLHQVN QAMMSNLTRQ GSPDCVIPFL PLESSPAQLS SDTASLLSGL VRLDEHSQIF ARKVANTFKP HRLQARKAMW RKEQDTRALR EQSQHLDGER MQAAALNAAY SAYLQSYLSY QAQMEQLQVA FGSHMSFGTG APYGARMPFG GQVPLGAPPP FPTWPGCPQP PPLHAWQAGT PPPPSPQPAA FPQSLPFPQS PAFPTASPAP PQSPGLQPLI IHHAQMVQLG LNNHMWNQRG SQAPEDKTQE AE | |
Greater than 85% as determined by SDS-PAGE. |
148022 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |