missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thrombopoietin / THPO Protein
A DNA sequence encoding the Human Thrombopoietin / THPO, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10387-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000590.1 | |
Thrombopoietin / THPO Protein | |
Research Use Only | |
IGFBP5 | |
The ED50 as determined by its ability to inhibit IGF-II induced proliferation of MCF-7 cells is less than 0.4 ug/ml, corresponding to a specific activity of > 2500 IU/mg in the presence of 15 ng/ml of rHuIGF-II. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.0, lyophilized |
3488 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LGSFVHCEPC DEKALSMCPP SPLGCELVKE PGCGCCMTCA LAEGQSCGVY TERCAQGLRC LPRQDEEKPL HALLHGRGVC LNEKSYREQV KIERDSREHE EPTTSEMAEE TYSPKIFRPK HTRISELKAE AVKKDRRKKL TQSKFVGGAE NTAHPRIISA PEMRQESEQG PCRRHMEASL QELKASPRMV PRAVYLPNCD RKGFYKRKQC KPSRGRKRGI CWCVDKYGMK LPGMEYVDGD FQCHTFDSSN VE | |
Greater than 96% as determined by SDS-PAGE and HPLC. |