missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF9 Protein
A cDNA sequence encoding the TAF9 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13686-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
TAF9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLLPNILLTG TPGVGKTTLG KELASKSGLK YINVGDLARE EQLYDGYDEE YDCPILDEDR VVDELDNQMR EGGVIVDYHG CDFFPERWFH IVFVLRTDTN VLYERLETRG YNEKKLTDNI QCEIFQVLYE EATASYKEEI VHQLPSNKPE ELENNVDQIL KWIEQWIKDH NS | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TAF9 Human solution (0.5 mg/1ml) containing 20mM Trsi HCL pH 8.0, 1mM DTT and 10% glycerol. |