Learn More
enQuireBio™ Recombinant Human STX1A Protein
A cDNA sequence encoding the STX1A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13633-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
6804 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKDRTQELRT AKDSDDDDDV AVTVDRDRFM DEFFEQVEEI RGFIDKIAEN VEEVKRKHSA ILASPNPDEK TKEELEELMS DIKKTANKVR SKLKSIEQSI EQEEGLNRSS ADLRIRKTQH STLSRKFVEV MSEYNATQSD YRERCKGRIQ RQLEITGRTT TSEELEDMLE SGNPAIFASG IIMDSSISKQ ALSEIETRHS EIIKLENSIR ELHDMFMDMA MLVESQGEMI DRIEYNVEHA VDYVERAVSD TKKAVKYQSK ARRKK | |
Greater than 95.0% as determined by SDS-PAGE. |
STX1A Protein | |
Research Use Only | |
STX1A | |
Human | |
Untagged | |
STX1A protein solution (1 mg/ml) containing phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |