missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOD1 / Superoxide Dismutase Protein
A DNA sequence encoding the Human SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10159-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000445.1 | |
SOD1 / Superoxide Dismutase Protein | |
Research Use Only | |
sod1 | |
The potency per mg was determined by Pyrogallic Acid method and was found to be more than 1.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6647 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ATKAVCVLKG DGPVQGIINF EQKESNGPVK VWGSIKGLTE GLHGFHVHEF GDNTAGCTSA GPHFNPLSRK HGGPKDEERH VGDLGNVTAD KDGVADVSIE DSVISLSGDH CIIGRTLVVH EKADDLGKGG NEESTKTGNA GSRLACGVIG IAQ | |
Greater than 95% as determined by SDS-PAGE and HPLC. |