missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPA1 Protein
A cDNA sequence encoding the SNRPA1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13542-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SNRPA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVKLTAELIE QAAQYTNAVR DRELDLRGYK IPVIENLGAT LDQFDAIDFS DNEIRKLDGF PLLRRLKTLL VNNNRICRIG EGLDQALPCL TELILTNNSL VELGDLDPLA SLKSLTYLSI LRNPVTNKKH YRLYVIYKVP QVRVLDFQKV KLKERQEAEK MFKGKRGAQL AKDIARRSKT FNPGAGLPTD KKKGGPSPGD VEAIKNAIAN ASTLAEVERL KGLLQSGQIP GRERRSGPTD DGEEEMEEDT VTNGS | |
Greater than 80% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRPA1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.2M NaCl,2mM DTT and 0.1mM PMSF. |