missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNAPIN Protein
A cDNA sequence encoding the SNAPIN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13534-50ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SNAPIN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAGAGSAAVS GAGTPVAGPT GRDLFAEGLL EFLRPAVQQL DSHVHAVRES QVELREQIDN LATELCRINEDQKVALDLDP YVKKLLNARR RVVLVNNILQ NAQERLRRLN HSVAKETARR RAMLDSGIYP PGSPGK | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution contains 20mM Tris-HCl pH 8.0, 5mM DTT, 2mM EDTA, 0.2M NaCl, and 40% glycerol. |