missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small Nuclear Ribonucleoprotein, 70kDa Protein
A cDNA sequence encoding the Small Nuclear Ribonucleoprotein, 70kDa was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP14031-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
6625 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MTQFLPPNLL ALFAPRDPIP YLPPLEKLPH EKHHNQPYCG IAPYIREFED PRDAPPPTRA ETREERMERK RREKIERRQQ EVETELKMWD PHNDPNAQGD AFKTLFVARV NYDTTESKLR REFEVYGPIK RIHMVYSKRS GKPRGYAFIE YEHERDMHSA YKHADGKKID GRRVLVDVER GRTVKGWRPR RLGGGLGGTR RGGADVNIRH SGRDDTSRYD ERPGPSPLPH RDRDRDRERE RRERSRERDK ERERRRSRSR DRRRRSRSRD KEERRRSRER SKDKDRDRKR RSSRSRERAR RERERKEELR GGGGDMAEPS EAGDAPPDDG PPGELGPDGP DGPEEKGRDR DRERRRSHRS ERERRRDRDR DRDRDREHKR GERGSERGRD EARGGGGGQD NGLEGLGNDS RDMYMESEGG DGYLAPENGY LMEAAPE | |
Greater than 90% as determined by SDS-PAGE. |
Small Nuclear Ribonucleoprotein, 70kDa Protein | |
Research Use Only | |
SNRNP70 | |
Human | |
His | |
U1-snRNP is supplied in 20mM HEPES buffer pH 7.5, 0.01mM EDTA and 0.02% SDS. |