missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH3BGRL2 Protein
A cDNA sequence encoding the SH3BGRL2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13488-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SH3BGRL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVIRVFI ASSSGFVAIK KKQQDVVRFL EANKIEFEEV DITMSEEQRQ WMYKNVPPEK KPTQGNPLPP QIFNGDRYCG DYDSFFESKE SNTVFSFLGL KPRLASKAEP | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SH3BGRL2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |