missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH3BGRL2 Protein
A cDNA sequence encoding the SH3BGRL2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Name | SH3BGRL2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15993729
|
enQuireBio™
QP13488-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15983729
|
enQuireBio™
QP13488-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15973729
|
enQuireBio™
QP13488-1MG |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
SH3BGRL2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SH3BGRL2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVIRVFI ASSSGFVAIK KKQQDVVRFL EANKIEFEEV DITMSEEQRQ WMYKNVPPEK KPTQGNPLPP QIFNGDRYCG DYDSFFESKE SNTVFSFLGL KPRLASKAEP | |
Greater than 90% as determined by SDS-PAGE. |