missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinI1 / Neuroserpin Protein
A DNA sequence encoding the Human SerpinI1 / Neuroserpin, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10160-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001116224.1 | |
SerpinI1 / Neuroserpin Protein | |
Research Use Only | |
SERPINI1 | |
The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 0.5 ug/ml, corresponding to a specific activity of > 2000 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.5, lyophilized |
5274 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TGATFPEEAI ADLSVNMYNR LRATGEDENI LFSPLSIALA MGMMELGAQG STQKEIRHS MGYDSLKNGE EFSFLKEFSN MVTAKESQYV MKIANSLFVQ NGFHVNEEF LQMMKKYFNA AVNHVDFSQN VAVANYINKW VENNTNNLVK DLVSPRDFD AATYLALINA VYFKGNWKSQ FRPENTRTFS FTKDDESEVQ IPMMYQQGE FYYGEFSDGS NEAGGIYQVL EIPYEGDEIS MMLVLSRQEV PLATLEPLV KAQLVEEWAN SVKKQKVEVY LPRFTVEQEI DLKDVLKALG ITEIFIKDA NLTGLSDNKE IFLSKAIHKS FLEVNEEGSE AAAVSGMIAI SRMAVLYPQ VIVDHPFFFL IRNRRTGTIL FMGRVMHPET MNTSGHD | |
Greater than 95% as determined by SDS-PAGE and HPLC. |