missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINA1 Protein
A cDNA sequence encoding the SERPINA1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10864-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SERPINA1 Protein | |
Research Use Only | |
3∼5 mg SERPINA1 will inhibit 1 mg PPE with an activity of 10.8 units per mg protein. | |
Human | |
Untagged | |
SERPINA1 was lyophilized from a concentrated solution containing recombinant Albumin. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Rice | |
MPSSVSWGIL LLAGLCCLVP VSLAEDPQGD AAQKTDTSHH DQDHPTFNKI TPNLAEFAFS LYRQLAHQSN STNIFFSPVS IATAFAMLSL GTKADTHDEI LEGLNFNLTE IPEAQIHEGF QELLRTLNQP DSQLQLTTGN GLFLSEGLKL VDKFLEDVKK LYHSEAFTVN FGDTEEAKKQ INDYVEKGTQ GKIVDLVKEL DRDTVFALVN YIFFKGKWER PFEVKDTEEE DFHVDQVTTV KVPMMKRLGM FNIQHCKKLS SWVLLMKYLG NATAIFFLPD EGKLQHLENE LTHDIITKFL ENEDRRSASL HLPKLSITGT YDLKSVLGQL GITKVFSNGA DLSGVTEEAP LKLSKAVHKA VLTIDEKGTE AAGAMFLEAI PMSIPPEVKF NKPFVFLMIE QNTKSPLFMG KVVNPTQK | |
Greater than 95% as determined by SDS-PAGE. |