missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAP18 Protein
A cDNA sequence encoding the SAP18 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13413-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SAP18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAVESRVTQE EIKKEPEKPI DREKTCPLLL RVFTTNNGRH HRMDEFSRGN VPSSELQIYT WMDATLKELT SLVKEVYPEA RKKGTHFNFA IVFTDVKRPG YRVKEIGSTM SGRKGTDDSM TLQSQKFQIG DYLDIAITPP NRAPPPSGRM RPY | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SAP18 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol, 1mM DTT. |