missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAA4 Protein
A cDNA sequence encoding the SAA4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13410-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
6291 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MWRSFFKEAL QGVGDMGRAY WDIMISNHQN SNRYLYARGN YDAAQRGPGG VWAAKLISRS RVYLQGLIDY YLFGNSSTVL EDSKSNEKAE EWGRSGKDPD RFRPDGLPKK Y | |
Greater than 90.0% as determined by SDS-PAGE. |
SAA4 Protein | |
Research Use Only | |
SAA4 | |
Human | |
His | |
The SAA4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 5mM DTT, 0.2M NaCl and 0.5mM EDTA. |