missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAA4 Protein
A cDNA sequence encoding the SAA4 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Gen-ID (Entrez) | 6291 |
---|---|
Name | SAA4 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | SAA4 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15993479
|
enQuireBio™
QP13410-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15973479
|
enQuireBio™
QP13410-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15983479
|
enQuireBio™
QP13410-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
6291 | |
Research Use Only | |
SAA4 | |
Human | |
His | |
The SAA4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 5mM DTT, 0.2M NaCl and 0.5mM EDTA. |
SAA4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MWRSFFKEAL QGVGDMGRAY WDIMISNHQN SNRYLYARGN YDAAQRGPGG VWAAKLISRS RVYLQGLIDY YLFGNSSTVL EDSKSNEKAE EWGRSGKDPD RFRPDGLPKK Y | |
Greater than 90.0% as determined by SDS-PAGE. |