missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100G Protein
A cDNA sequence encoding the S100G was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13407-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
S100G Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS TKKSPEELKRS TKKSPEELKR IFEKYAAKEG DPDQLSKDEL KLLIQAEFPS LLKGPNTLDD LFQELDKNGD GEVSFEEFQV LVKKISQ |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
S100G was filtered (0.4 microm) and lyophilized in 0.5 mg/ml in 20mM Tris and 50mM NaCl, pH 7.5. |