missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RYBP Protein
A cDNA sequence encoding the RYBP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13379-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
23429 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTMGDK KSPTRPKRQA KPAADEGFWD CSVCTFRNSA EAFKCSICDV RKGTSTRKPR INSQLVAQQV AQQYATPPPP KKEKKEKVEK QDKEKPEKDK EISPSVTKKN TNKKTKPKSD ILKDPPSEAN SIQSANATTK TSETNHTSRP RLKNVDRSTA QQLAVTVGNV TVIITDFKEK TRSSSTSSST VTSSAGSEQQ NQSSSGSEST DKGSSRSSTP KGDMSAVNDE SF | |
Greater than 85% as determined by SDS-PAGE. |
RYBP Protein | |
Research Use Only | |
RYBP | |
Human | |
His | |
The RYBP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |