missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHOV Protein
A cDNA sequence encoding the RHOV was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13320-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
RHOV Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPPRELS EAEPPPLRAP TPPPRRRSAP PELGIKCVLV GDGAVGKSSL IVSYTCNGYP ARYRPTALDT FSVQVLVDGA PVRIELWDTA GQEDFDRLRS LCYPDTDVFL ACFSVVQPSS FQNITEKWLP EIRTHNPQAP VLLVGTQADL RDDVNVLIQL DQGGREGPVP QPQAQGLAEK IRACCYLECS ALTQKNLKEV FDSAILSAIE HKARLEKKLN AKGVRTLSRC RWKKFFCFV | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RHOV solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 2M Urea, 1mM DTT and 20% glycerol. |