missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGS14 Protein
A cDNA sequence encoding the RGS14 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13307-100ug
Additional Details : Gewicht : 0.01000kg
Specifications
10636 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPGKPKHLGV PNGRMVLAVS DGELSSTTGP QGQGEGRGSS LSIHSLPSGP SSPFPTEEQP VASWALSFER LLQDPLGLAY FTEFLKKEFS AENVTFWKAC ERFQQIPASD TQQLAQEARN IYQEFLSSQA LSPVNIDRQA WLGEEVLAEP RPDMFRAQQL QIFNLMKFDS YARFVKSPLY RECLLAEAEG RPLREPGSSR LGSPDATRKK PKLKPGKSLP LGVEELGQLP PVEGPGGRPL RKSFRRELGG TANAALRRES QGSLNSSASL DLGFLAFVSS KSESHRKSLG STEGESESRP GKYCCVYLPD GTASLALARP GLTIRDMLAG ICEKRGLSLPDIKVYLVGNE QALVLDQDCT VLADQEVRLE NRITFELELT ALERVVRISA KPTKRLQEAL QPILEKHGLS PLEVVLHRPG EKQPLDLGKL VSSVAAQRLV LDTLPGVKIS KARDKSPCRS QGCPPRTQDK ATHPPPASPS SLVKVPSSAT GKRQTCDIEG LVELLNRVQS SGAHDQRGLLRKEDLVLPEF LQLPAQGPSS EETPPQTKSA AQPIGGSLNS TTDSAL | |
Greater than 80% as determined by SDS-PAGE. |
RGS14 Protein | |
Research Use Only | |
RGS14 | |
Human | |
His | |
RGS14 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 7.5, 10% glycerol, 1mM DTT and 200mM NaCl. |