missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RGS1 Protein
A cDNA sequence encoding the RGS1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13306-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
RGS1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMRAAAI STPKLDKMPG MFFSANPKEL KGTTHSLLDD KMQKRRPKTF GMDMKAYLRS MIPHLESGMK SSKSKDVLSA AEVMQWSQSL EKLLANQTGQ NVFGSFLKSE FSEENIEFWL ACEDYKKTES DLLPCKAEEI YKAFVHSDAA KQINIDFRTR ESTAKKIKAP TPTCFDEAQK VIYTLMEKDS YPRFLKSDIY LNLLNDLQAN SLK | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RGS1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |